Skip to Content
Merck
Hydrogen peroxide 100 vol univAR™, 30%

Hydrogen peroxide 100 vol

Hydrogen peroxide 100 vol univAR™, 30%
Synonym(s):
Hydrogen peroxide solution
Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01

Products

Formula Weight
Product Line
Markush Class
Markush Group
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing