Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hi-Flow
™
Plus Membrane 180
Products
Product Category
membranes (12)
rolls (7)
sheets (5)
Brand
Millipore (12)
Length (m)
Quality Segment
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
HF180MC100
Hi-Flow
™
Plus Membrane 180,
size 6 cm × 30.1 cm, pack of 100 laminated membrane cards
Expand
View Pricing
HF180MC5PK
Hi-Flow
™
Plus Membrane 180,
size 6 cm × 30.1 cm, pack of 5 laminated membrane cards
Expand
View Pricing
SHF1800425
Hi-Flow
™
Plus Membrane 180,
backing thickness 4 mil, width 25 mm, pack of 1 slit roll
Expand
View Pricing
HF180UBXSS
Hi-Flow
™
Plus Membrane 180,
size 30.5 cm × 25 cm , unbacked, pack of 5 sheets
Expand
View Pricing
HF18002XSS
Hi-Flow
™
Plus Membrane 180,
backing thickness 2 mil, size 30.5 cm × 25 cm, pack of 5 sheets
Expand
View Pricing
SHF180UB25
Hi-Flow
™
Plus Membrane 180,
width 25 mm , unbacked, pack of 1 slit roll
Expand
View Pricing
SHF1800225
Hi-Flow
™
Plus Membrane 180,
backing thickness 2 mil, width 25 mm, pack of 1 slit roll
Expand
View Pricing
BPCKHF18004S25
Hi-Flow
™
Plus Membrane 180,
4mil backing, 25mm width, pack of 11 slit rolls
Expand
View Pricing
HF18004XSS
Hi-Flow
™
Plus Membrane 180,
backing thickness 4 mil, size 30.5 cm × 25 cm, pack of 5 sheets
Expand
View Pricing
BPCKHF18004S20
Hi-Flow
™
Plus Membrane 180,
4mil backing, 20mm width, pack of 14 slit rolls
Expand
View Pricing
BPCKHF18002S20
Hi-Flow
™
Plus Membrane 180,
2mil backing, 20mm width, pack of 14 slit rolls
Expand
View Pricing
BPCKHF18002S25
Hi-Flow
™
Plus Membrane 180,
2mil backing, 25mm width, pack of 11 slit rolls
Expand
View Pricing