Skip to Content
Merck
Hexylamine analytical standard

Hexylamine

Hexylamine analytical standard
Synonym(s):
1-Aminohexane, Hexylamine
Linear Formula:
CH3(CH2)5NH2
CAS Number:
Molecular Weight:
101.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing