Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hexamethylene diisocyanate
Synonym(s):
1,6-Diisocyanatohexane, Hexamethylene diisocyanate
Linear Formula:
OCN(CH
2
)
6
NCO
CAS Number:
822-06-0
Molecular Weight:
168.19
EC Number:
212-485-8
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Puriss (1)
Purum (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
52649
Hexamethylene diisocyanate,
puriss., ≥99.0% (GC)
Expand
View Pricing
8.22066
Hexamethylene diisocyanate,
for synthesis
Expand
View Pricing
52650
Hexamethylene diisocyanate,
purum, ≥98.0% (GC)
Expand
View Pricing