
Heptachlor endo-epoxide

Synonym(s):
endo-1,4,5,6,7,8,8-Heptachloro-2,3-epoxy-4,7-methano-3a,4,7,7a-tetrahydroindane
Empirical Formula (Hill Notation):
C10H5Cl7O
CAS Number:
Molecular Weight:
389.32
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing