
Haloperidol decanoate

Synonym(s):
Decanoic acid 4-(4-chlorophenyl)-1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidinyl ester, Haldol decanoate
Empirical Formula (Hill Notation):
C31H41ClFNO3
CAS Number:
Molecular Weight:
530.11
Products
Color
Feature
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3