Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gold-germanium eutectic alloy, Au88Ge12
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF36153705
Gold-germanium eutectic alloy, Au88Ge12,
5g lump, 10mm max. lump size
Expand
View Pricing
GF58689220
Gold-germanium eutectic alloy, Au88Ge12,
2g lump, 10mm max. lump size
Expand
View Pricing
GF43865196
Gold-germanium eutectic alloy, Au88Ge12,
foil, 5m coil, 1mm coil width, 0.025mm thickness
Expand
View Pricing
GF08858087
Gold-germanium eutectic alloy, Au88Ge12,
foil, 0.2m coil, 1mm coil width, 0.025mm thickness
Expand
View Pricing
GF07260454
Gold-germanium eutectic alloy, Au88Ge12,
foil, 0.5m coil, 1mm coil width, 0.025mm thickness
Expand
View Pricing
GF87874802
Gold-germanium eutectic alloy, Au88Ge12,
foil, 1m coil, 1mm coil width, 0.025mm thickness
Expand
View Pricing