Skip to Content
Merck
Glycycoumarin ≥90% (LC/MS-ELSD)

Glycycoumarin

Glycycoumarin ≥90% (LC/MS-ELSD)
Empirical Formula (Hill Notation):
C21H20O6
CAS Number:
Molecular Weight:
368.38

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing