
Glycopyrronium bromide

Synonym(s):
Glycopyrrolate, α-cyclopentylmandelic acid ester with 3-hydroxy-1,1-dimethylpyrrolidinium bromide, 1-methyl-3-pyrrolidyl α-cyclopentylmandelate methobromide, 1-methyl-3-pyrrolidyl α-phenyl-α-cyclopentylglycolate methobromide, 3-(2-phenyl-2-cyclopentylglycoloyloxy)-1,1-dimethylpyrrolidinium bromide, 3-hydroxy-1,1-dimethylpyrrolidinium bromide α-cyclopentylmandelate, AHR-504, Copyrrolate, Gastrodyn, NSC 250836, NSC 251251, NSC 251252, NVA-237, glycopyrronium bromide
Empirical Formula (Hill Notation):
C19H28NO3 · Br
CAS Number:
Molecular Weight:
398.33
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing