
Glyceryl distearate

Synonym(s):
Distearoylglycerol mixed isomers
Empirical Formula (Hill Notation):
C39H76O5
CAS Number:
Molecular Weight:
625.02
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing