Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glucose Oxidase from
Aspergillus niger
Synonym(s):
β-D-Glucose:oxygen 1-oxidoreductase, G.Od., GOx
CAS Number:
9001-37-0
EC Number:
232-601-0
Products
Brand
Sigma-Aldrich (2)
Sustainability
Feature
Physical Form
Chemical Composition
Quality Segment
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
G7141
Glucose Oxidase from
Aspergillus niger
,
Type X-S, lyophilized powder, 100,000-250,000 units/g solid (without added oxygen)
Expand
View Pricing
G2133
Glucose Oxidase from
Aspergillus niger
,
Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Expand
View Pricing