Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glass Fiber Diagnostic Pad
Products
Product Category
pads (7)
sheets (3)
rolls (2)
strips (2)
Brand
Millipore (7)
Length (cm)
Length (m)
Quality Segment
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
GFDX083000
Glass Fiber Diagnostic Pad,
filter W × L 8 mm × 300 mm, pack of 100 strips
Expand
View Pricing
GFDX103000
Glass Fiber Diagnostic Pad,
filter W × L 10 mm × 300 mm, pack of 100 strips
Expand
View Pricing
GFDX203000
Glass Fiber Diagnostic Pad,
filter L × W 20 cm × 30 cm, pack of 100 sheets
Expand
View Pricing
GFDX20305PK
Glass Fiber Diagnostic Pad,
filter L × W 20 cm × 30 cm , Pad Size, pack of 5 sheets
Expand
View Pricing
GFDX001000
Glass Fiber Diagnostic Pad,
filter W × L 10 mm × 100 m, pack of 1 roll
Expand
View Pricing
GFDX000800
Glass Fiber Diagnostic Pad,
filter W × L 8 mm × 100 m, pack of 1 roll
Expand
View Pricing
GFDX202500
Glass Fiber Diagnostic Pad,
filter L × W 20.32 cm × 25.4 cm, pack of 100 sheets
Expand
View Pricing