Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Furaltadone
Synonym(s):
5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidinone
Empirical Formula (Hill Notation):
C
13
H
16
N
4
O
6
CAS Number:
139-91-3
Molecular Weight:
324.29
EC Number:
205-384-5
Products
Product Category
antibiotics (1)
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Product Line
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
F9130
Furaltadone,
Expand
View Pricing
46289
Furaltadone,
VETRANAL
®
, analytical standard
Expand
View Pricing