Skip to Content
Merck
Fractogel® EMD COO- (M)

Fractogel® EMD COO- (M)

Fractogel® EMD COO- (M)

Products

Product Category
Brand
Physical Form
Product Line
Analyte Suitability
Separation Technique
Quality Segment
Application
Particle Size (µm)
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing