Skip to Content
Merck
Formamide solution NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.

Formamide solution

Formamide solution NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Empirical Formula (Hill Notation):
CH3NO
Molecular Weight:
45.04

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Technique