
Formamide, Molecular Biology Grade

Synonym(s):
Formamide, Amide C1, Formic amide
Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing