
Fmoc-Arg(Pbf)-OH

Synonym(s):
Fmoc-Arg(Pbf)-OH, Nα-Fmoc-Nω-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine, Nα-Fmoc-Nω-Pbf-L-arginine
Empirical Formula (Hill Notation):
C34H40N4O7S
CAS Number:
Molecular Weight:
648.77
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing