
Fluvoxamine impurity D

Synonym(s):
5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one
Empirical Formula (Hill Notation):
C13H15F3O2
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing