
Flutolanil

Synonym(s):
N-(3-Isopropoxyphenyl)-2-(trifluoromethyl)benzamide
Empirical Formula (Hill Notation):
C17H16F3NO2
CAS Number:
Molecular Weight:
323.31
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing