
Flunixin impurity B

Synonym(s):
2-Methyl-3-(trifluoromethyl)aniline, 2-Amino-6-(trifluoromethyl)toluene
Linear Formula:
CH3C6H3(CF3)NH2
CAS Number:
Molecular Weight:
175.15
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing