Skip to Content
Merck
Fentanyl related compound G certified reference material, pharmaceutical secondary standard

Fentanyl Related Compound G

Fentanyl related compound G certified reference material, pharmaceutical secondary standard
Synonym(s):
Acetyl fentanyl, N-Phenyl-N-[1-(2-phenylethyl)-4-piperidinyl]-acetamide
Empirical Formula (Hill Notation):
C21H26N2O
CAS Number:
Molecular Weight:
322.44

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade