Skip to Content
Merck
Famciclovir Pharmaceutical Secondary Standard; Certified Reference Material

Famciclovir

Famciclovir Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-(2-(2-Amino-9H-purin-9-yl)ethyl)-1,3-propanediol diacetate ester, BRL 42810, Famciclovir
Empirical Formula (Hill Notation):
C14H19N5O4
CAS Number:
Molecular Weight:
321.33

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing