
Ethyl viologen diiodide

Synonym(s):
1,1′-Diethyl-4,4′-bipyridinium diiodide
Empirical Formula (Hill Notation):
C14H18I2N2
CAS Number:
Molecular Weight:
468.12
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing