Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ethyl
trans
-2-butenoate
Synonym(s):
Ethyl crotonate
Linear Formula:
CH
3
CH=CHCOOC
2
H
5
CAS Number:
623-70-1
EC Number:
210-808-7
Products
Product Category
aroma chemicals (2)
Brand
Sigma-Aldrich (2)
Biological Source
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Food Grade (2)
Kosher (2)
Halal (1)
Natural (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
W348600
Ethyl
trans
-2-butenoate,
≥96%, FG
Expand
View Pricing
W348630
Ethyl
trans
-2-butenoate,
natural, ≥97%, FG
Expand
View Pricing