Ethylene sulfide
Synonym(s):
Dimethylene sulfide, Thiirane
Empirical Formula (Hill Notation):
C2H4S
CAS Number:
Molecular Weight:
60.12
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing