
Ethylene glycol 5 M solution

Synonym(s):
Ethylene glycol solution
Empirical Formula (Hill Notation):
C2H6O2
CAS Number:
Molecular Weight:
62.07
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing