Skip to Content
Merck
Ethanol-water solution NIST® SRM® 2895, nominal mass fraction 0.2%

Ethanol-water solution

Ethanol-water solution NIST® SRM® 2895, nominal mass fraction 0.2%
Synonym(s):
Ethanol solution, Ethyl alcohol
Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07

Products

Formula Weight
Manufacturer
Format
Quality Segment
Application
Special Grade
Technique