Ethacrynic acid
Synonym(s):
(2,3-Dichloro-4-[2-methylenebutyryl]phenoxy)acetic acid
Empirical Formula (Hill Notation):
C13H12Cl2O4
CAS Number:
Molecular Weight:
303.14
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3