
Estrone 3-(β-D-glucuronide) sodium salt

Synonym(s):
1,3,5(10)-Estratrien-17-one 3-glucuronide sodium salt
Empirical Formula (Hill Notation):
C24H29NaO8
CAS Number:
Molecular Weight:
468.47
Products
Product Category
Brand
Formula Weight
Physical Form
Research Area
Quality Segment
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing