
Estriol

Synonym(s):
1,3,5(10)-Estratriene-3,16α,17β-triol, 16α-Hydroxyestradiol, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene
Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility
Showing 1-5 of 5