Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Estapor
®
White Microspheres
Products
Brand
Sigma-Aldrich (4)
Color
Quality Segment
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
FR123690087
Estapor
®
White Microspheres,
K-030 White Std PS 10%, White microspheres
300nm
, Latex Turbidimetric ImmunAssay - For ordering, click "Request more information"
Expand
View Pricing
FR139430087
Estapor
®
White Microspheres,
K-020 White Std PS 10%, White microspheres
200nm
, Latex Turbidimetric Immunoassay - For ordering, click "Request more information"
Expand
View Pricing
FR139469081
Estapor
®
White Microspheres,
K-010 White Std PS 10%, White microspheres
100nm
, Latex Turbidimetric Immunoassay - For ordering, click "Request more information"
Expand
View Pricing
FR139431081
Estapor
®
White Microspheres,
K-007 White Std PS 10%, White microspheres
70nm
, Latex Turbidimetric Immunoassay
Expand
View Pricing