Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Estapor
®
Fluorescent Carboxylated microspheres
Products
Brand
Sigma-Aldrich (3)
Color
Quality Segment
Functional Group
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
FR180380624
Estapor
®
Fluorescent Carboxylated microspheres,
F1-EU-030 FluoEU
COOH
PS 1%,
Red
Fluorescent carboxylated microspheres
300nm
, Lateral Flow Immunoassay - For ordering, click "Request more information"
Expand
View Pricing
FR180380623
Estapor
®
Fluorescent Carboxylated microspheres,
F1-EU-010 FluoEU
COOH
PS 1%,
Red
Fluorescent carboxylated microspheres
160nm
, Lateral Flow Immunoassay - For ordering, click "Request more information"
Expand
View Pricing
FR180380625
Estapor
®
Fluorescent Carboxylated microspheres,
F1-EU-050 FluorEU
COOH
PS 1%,
Red
Fluorescent carboxylated microspheres
500nm
, Lateral Flow Immunoassay - For ordering, click "Request more information"
Expand
View Pricing