Skip to Content
Merck
Esomeprazole Magnesium Pharmaceutical Secondary Standard; Certified Reference Material

Esomeprazole magnesium

Esomeprazole Magnesium Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(S)-Omeprazole magnesium trihydrate
Empirical Formula (Hill Notation):
C34H36MgN6O6S2 · 3H2O
CAS Number:
Molecular Weight:
767.17

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique