Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Erucamide
Synonym(s):
cis
-13-Docosenoamide, Erucamide
Linear Formula:
CH
3
(CH
2
)
7
CH=CH(CH
2
)
11
CONH
2
CAS Number:
112-84-5
Molecular Weight:
337.58
EC Number:
204-009-2
Products
Product Category
lipids (1)
Brand
Supelco (1)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
90082
Erucamide,
analytical standard
Expand
View Pricing