Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Replacement channel for multichannel lower parts
Products
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
EP4861604000
Eppendorf
®
Replacement channel for multichannel lower parts,
size 1200 μL, green
Expand
View Pricing
EP4922609000
Eppendorf
®
Replacement channel for multichannel lower parts,
size 300 μL, orange, includes piston, cylinder, tip spring, o ring
Expand
View Pricing
EP4922607007
Eppendorf
®
Replacement channel for multichannel lower parts,
size 10 μL, medium gray, includes piston, cylinder, tip spring
Expand
View Pricing
EP4922608003
Eppendorf
®
Replacement channel for multichannel lower parts,
size 100 μL, yellow, includes piston, cylinder, tip spring, o ring
Expand
View Pricing