Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Pipette lower part
Products
Product Category
pipettors (4)
Color
Manufacturer
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
EP4861603003
Eppendorf
®
Pipette lower part,
single channel, size 1-10 mL, turquoise
Expand
View Pricing
EP4861607009
Eppendorf
®
Pipette lower part,
12-channel, size 0.5-10 μL, gray
Expand
View Pricing
EP4861610000
Eppendorf
®
Pipette lower part,
8-channel, size 15-300 μL, orange
Expand
View Pricing
EP4861609001
Eppendorf
®
Pipette lower part,
12-channel, size 5-100 μL, yellow
Expand
View Pricing