Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Microplate 96/F-PP
Products
Product Category
plates (3)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Capacity (mL)
Wells
Color
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
EP0030601475
Eppendorf
®
Microplate 96/F-PP,
sterile (PCR clean), flat bottom white wells, white wells, gray border, pkg of 80 plates (5 bags x 16 plates)
Expand
View Pricing
EP0030601700
Eppendorf
®
Microplate 96/F-PP,
PCR clean, flat bottom, black wells, white border, pkg of 80 ea (5 bags of 16 plates)
Expand
View Pricing
EP0030602102
Eppendorf
®
Microplate 96/F-PP,
clear wells, white border, sterile
Expand
View Pricing