
Epidermal Growth Factor from murine submaxillary gland

Synonym(s):
EGF, Epidermal cell growth factor
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Color
Gene Alias
Physical Form
Product Line
Tissue
Quality Segment
Technique
Showing 1-2 of 2