Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
EPA 8270 Semivolatile Internal Standard Mix
Products
Brand
Supelco (5)
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
certified reference material (5)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CRM46955
EPA 8270 Semivolatile Internal Standard Mix,
certified reference material,
Trace
CERT
®
, 2000 μg/mL each component in methylene chloride, ampule of 1 mL
Expand
View Pricing
CRM48902
EPA 8270 Semivolatile Internal Standard Mix,
certified reference material,
Trace
CERT
®
, 2000 μg/mL each component in methylene chloride, ampule of 2 × 1 mL
Expand
View Pricing
CRM47906
EPA 8270 Semivolatile Internal Standard Mix,
High Concentration, certified reference material, 4000 μg/mL each component in methylene chloride: benzene (1:1), ampule of 1 mL
Expand
View Pricing
CRM5M07296
EPA 8270 Semivolatile Internal Standard Mix,
certified reference material,
Trace
CERT
®
, 2000 μg/mL each component in dichloromethane, ampule of 5 × 1 mL
Expand
View Pricing
46866-U
EPA 8270 Semivolatile Internal Standard Mix,
certified reference material, 2000 μg/mL each component in methylene chloride, ampule of 25 mL
Expand
View Pricing