
Ebselen

Synonym(s):
2-Phenyl-1,2-benzisoselenazol-3(2H)-one, Ebselen
Empirical Formula (Hill Notation):
C13H9NOSe
CAS Number:
Molecular Weight:
274.18
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing