Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
Measuring Cylinder
Products
Product Category
cylinders (22)
Capacity (mL)
Glassware Class
Color
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
DWK213905408
Duran
®
Measuring Cylinder,
graduated, MAIN POINTS RING GRADUATION, class A, hexagonal blue glass cylinder (base), cylinder capacity 1000 mL
Expand
View Pricing
DWK213972409
Duran
®
Measuring Cylinder,
cylinder capacity 100 mL, class A, UKAS CERTIFICATE
Expand
View Pricing
DWK213965405
Duran
®
Measuring Cylinder,
hexagonal glass cylinder (base), class B, graduated, WHITE GRADUATION, cylinder capacity 1000 mL
Expand
View Pricing
DWK213962408
Duran
®
Measuring Cylinder,
hexagonal glass cylinder (base), class B, graduated, WHITE GRADUATION, cylinder capacity 100 mL
Expand
View Pricing
DWK213975406
Duran
®
Measuring Cylinder,
cylinder capacity 1000 mL, class A, UKAS CERTIFICATE
Expand
View Pricing
DWK213974401
Duran
®
Measuring Cylinder,
cylinder capacity 500 mL, class A, UKAS CERTIFICATE
Expand
View Pricing
DWK213963601
Duran
®
Measuring Cylinder,
hexagonal glass cylinder (base), class B, graduated, WHITE GRADUATION, cylinder capacity 250 mL
Expand
View Pricing
DWK213973602
Duran
®
Measuring Cylinder,
cylinder capacity 250 mL, class A, UKAS CERTIFICATE
Expand
View Pricing
DWK213964409
Duran
®
Measuring Cylinder,
hexagonal glass cylinder (base), class B, graduated, WHITE GRADUATION, cylinder capacity 500 mL
Expand
View Pricing
DWK213906301
Duran
®
Measuring Cylinder,
graduated, MAIN POINTS RING GRADUATION, class A, hexagonal blue glass cylinder (base), cylinder capacity 2000 mL
Expand
View Pricing
DWK213961703
Duran
®
Measuring Cylinder,
hexagonal glass cylinder (base), class B, graduated, WHITE GRADUATION, cylinder capacity 50 mL
Expand
View Pricing
DWK213961403
Duran
®
Measuring Cylinder,
hexagonal glass cylinder (base), class B, graduated, WHITE GRADUATION, cylinder capacity 25 mL
Expand
View Pricing
DWK213900701
Duran
®
Measuring Cylinder,
graduated, MAIN POINTS RING GRADUATION, class A, hexagonal blue glass cylinder (base), cylinder capacity 5 mL
Expand
View Pricing
DWK213900804
Duran
®
Measuring Cylinder,
graduated, MAIN POINTS RING GRADUATION, class A, hexagonal blue glass cylinder (base), cylinder capacity 10 mL
Expand
View Pricing
DWK213966307
Duran
®
Measuring Cylinder,
hexagonal glass cylinder (base), class B, graduated, WHITE GRADUATION, cylinder capacity 2000 mL
Expand
View Pricing
DWK213976308
Duran
®
Measuring Cylinder,
cylinder capacity 2000 mL, class A, UKAS CERTIFICATE
Expand
View Pricing
DWK213970708
Duran
®
Measuring Cylinder,
cylinder capacity 5 mL, class A, UKAS CERTIFICATE
Expand
View Pricing
DWK213970802
Duran
®
Measuring Cylinder,
cylinder capacity 10 mL, class A, UKAS CERTIFICATE
Expand
View Pricing
DWK213971404
Duran
®
Measuring Cylinder,
cylinder capacity 25 mL, class A, UKAS CERTIFICATE
Expand
View Pricing
DWK213960707
Duran
®
Measuring Cylinder,
hexagonal glass cylinder (base), class B, graduated, WHITE GRADUATION, cylinder capacity 5 mL
Expand
View Pricing
Page 1 of 2
Page 1 of 2