Dodecylethyldimethylammonium bromide
Synonym(s):
Dimethyldodecylethylammonium bromide, Dodecyldimethylethylammonium bromide
Linear Formula:
[CH3(CH2)11N+(CH3)2C2H5]Br-
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1