
Docosanol (n-Docosanol)

Synonym(s):
1-Docosanol, Behenyl alcohol
Linear Formula:
CH3(CH2)21OH
CAS Number:
Molecular Weight:
326.60
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing