
DL-Buthionine-(S,R)-sulfoximine

Linear Formula:
CH3(CH2)3S(O)(=NH)CH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
222.31
Products
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing