Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(±)-α-Terpinyl acetate, predominantly α-isomer
Synonym(s):
(±)-2-(4-Methyl-3-cyclohexenyl)isopropyl acetate
Empirical Formula (Hill Notation):
C
12
H
20
O
2
CAS Number:
80-26-2
Molecular Weight:
196.29
EC Number:
201-265-7
Products
Brand
Supelco (1)
Boiling Point (°C)
Formula Weight
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
09206
(±)-α-Terpinyl acetate, predominantly α-isomer,
analytical standard
Expand
View Pricing