Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DL-alpha-Tocopherol acetate
Synonym(s):
Vitamin E acetate,
O
-Acetyl-α-tocopherol, DL-α-Tocopherol acetate, DL-α-Tocopheryl acetate
Empirical Formula (Hill Notation):
C
31
H
52
O
3
CAS Number:
7695-91-2
Molecular Weight:
472.74
EC Number:
231-710-0
Products
Product Category
vitamins (1)
Brand
Supelco (1)
Boiling Point (°C)
Formula Weight
Format
Quality Segment
Application
Optical Isomer
Package Size
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
47786
DL-alpha-Tocopherol acetate,
analytical standard
Expand
View Pricing