DL-5-Hydroxylysine hydrochloride
Synonym(s):
2,6-Diamino-5-hydroxycaproic acid hydrochloride, 2,6-Diamino-5-hydroxyhexanoic acid hydrochloride
Linear Formula:
NH2CH2CH(OH)CH2CH2CH(NH2)COOH · HCl
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Amino Acid
Showing 1-1 of 1