Skip to Content
Merck
Di-tert-butyl-iminodicarboxylate ≥96%

Di-tert-butyl-iminodicarboxylate

Di-tert-butyl-iminodicarboxylate ≥96%
Synonym(s):
N-Boc-tert-butylcarbamate, tert-Butyl iminodicarboxylate
Linear Formula:
[(CH3)3COCO]2NH
CAS Number:
Molecular Weight:
217.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing