Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Disperse Orange 3
Synonym(s):
4-(4-Nitrophenylazo)aniline
Linear Formula:
O
2
NC
6
H
4
N=NC
6
H
4
NH
2
CAS Number:
730-40-5
Molecular Weight:
242.23
EC Number:
211-984-8
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Melting Point (°C)
Purity
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
364797
Disperse Orange 3,
Dye content 90 %
Expand
View Pricing
53882
Disperse Orange 3,
analytical standard
Expand
View Pricing