Dipropyl phthalate
Linear Formula:
C6H4-1,2-(CO2CH2CH2CH3)2
CAS Number:
Molecular Weight:
250.29
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2